MAN182107789 Manchester Organics

4-Methylthio-3-(trifluoromethyl)aniline

  • MDL number MFCD04972923
  • Cas Number : 63094-56-4
  • Molecular Weight : 207.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(methylsulfanyl)-3-(trifluoromethyl)aniline
Inchi Code 1S/C8H8F3NS/c1-13-7-3-2-5(12)4-6(7)8(9,10)11/h2-4H,12H2,1H3
Inchi Key MBBRGNWOSWJCGG-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/X10683
Availability details for
Quantity
  • Availability
  • Add to Cart