MAN182107797 Manchester Organics

2-Amino-5-chloro-3-methylpyridine

  • MDL number MFCD00129030
  • Cas Number : 20712-16-7
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-3-methyl-2-pyridinylamine
Inchi Code 1S/C6H7ClN2/c1-4-2-5(7)3-9-6(4)8/h2-3H,1H3,(H2,8,9)
Inchi Key QTZBTBLHYPSFMG-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/A18047
Availability details for
Quantity
  • Availability
  • Add to Cart