MAN182107837 Manchester Organics

5-(3-Methoxy-phenyl)-oxazole

  • MDL number MFCD09749681
  • Molecular Weight : 175.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T18299

Safety Information

MSDS https://www.manchesterorganics.com/T18299
Availability details for
Quantity
  • Availability
  • Add to Cart