MAN182107855 Manchester Organics

N-Methyl-pyrazine-2,3-diamine

  • MDL number MFCD08460070
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E18145

Safety Information

MSDS https://www.manchesterorganics.com/E18145
Availability details for
Quantity
  • Availability
  • Add to Cart