MAN182107868 Manchester Organics

4-(3-Bromo-phenyl)-6-methyl-2-oxo-1,2,3,4-tetrahydro-pyrimidine-5-carboxylic acid ethyl ester

  • MDL number MFCD00176375
  • Molecular Weight : 339.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S18275

Safety Information

MSDS https://www.manchesterorganics.com/S18275
Availability details for
Quantity
  • Availability
  • Add to Cart