MAN182107912 Manchester Organics

2-Naphthalen-2-yl-piperazine

  • MDL number MFCD05864679
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A16318

Safety Information

MSDS https://www.manchesterorganics.com/A16318
Availability details for
Quantity
  • Availability
  • Add to Cart