MAN182107915 Manchester Organics

3-Bromo-2-fluorobenzonitrile

  • MDL number MFCD06657980
  • Molecular Weight : 200.008

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A18093
Availability details for
Quantity
  • Availability
  • Add to Cart