MAN182107919 Manchester Organics

1-(2-Nitro-phenyl)-3-phenyl-piperazine

  • MDL number MFCD07371418
  • Cas Number : 904818-42-4
  • Molecular Weight : 283.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/P16308
Availability details for
Quantity
  • Availability
  • Add to Cart