MAN182107933 Manchester Organics

5-Thiophen-3-yl-2H-pyrazole-3-carboxylic acid

  • MDL number MFCD04323087
  • Molecular Weight : 194.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R16499

Safety Information

MSDS https://www.manchesterorganics.com/R16499
Availability details for
Quantity
  • Availability
  • Add to Cart