MAN182107935 Manchester Organics

5-(2-Methoxy-phenyl)-2H-pyrazole-3-carboxylic acid methyl ester

  • MDL number MFCD03761321
  • Cas Number : 898052-17-0
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U16337
Availability details for
Quantity
  • Availability
  • Add to Cart