MAN182107958 Manchester Organics

4-Oxo-1,4-dihydro-[1,8]naphthyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD09749669
  • Molecular Weight : 218.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J18174

Safety Information

MSDS https://www.manchesterorganics.com/J18174
Availability details for
Quantity
  • Availability
  • Add to Cart