MAN182108010 Manchester Organics

2-Bromo-6-fluoroanisole

  • MDL number MFCD04974125
  • Cas Number : 845829-94-9
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/B16463
Availability details for
Quantity
  • Availability
  • Add to Cart