MAN182108066 Manchester Organics

2-Chloro-5-iodobenzotrifluoride

  • MDL number MFCD09037801
  • Cas Number : 260355-20-2
  • Molecular Weight : 306.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F16513
Availability details for
Quantity
  • Availability
  • Add to Cart