MAN182108116 Manchester Organics

2,5-Difluoro-4-hydroxybenzaldehyde

  • MDL number MFCD04112521
  • Cas Number : 918523-99-6
  • Molecular Weight : 158.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/H18196
Availability details for
Quantity
  • Availability
  • Add to Cart