MAN182108120 Manchester Organics

4-Bromo-2-chloro-1-(trifluoromethoxy)benzene

  • MDL number MFCD10699114
  • Molecular Weight : 275.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P18557

Safety Information

MSDS https://www.manchesterorganics.com/P18557
Availability details for
Quantity
  • Availability
  • Add to Cart