MAN182108124 Manchester Organics

1H-Indazole-4-carbaldehyde

  • MDL number MFCD06738279
  • Molecular Weight : 146.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/M18461
Availability details for
Quantity
  • Availability
  • Add to Cart