MAN182108128 Manchester Organics

3-Fluoro-2-methoxy-5-(trifluoromethyl)pyridine

  • MDL number MFCD10699120
  • Cas Number : 1138011-20-7
  • Molecular Weight : 195.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/N18485
Availability details for
Quantity
  • Availability
  • Add to Cart