MAN182108131 Manchester Organics

2,3-Dichloro-5-fluoropyridine

  • MDL number MFCD08669666
  • Molecular Weight : 165.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P18534
Availability details for
Quantity
  • Availability
  • Add to Cart