MAN182108140 Manchester Organics

2-Isopropyl-1H-benzoimidazole-5-carboxylic acid

  • MDL number MFCD03042800
  • Molecular Weight : 204.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E16654

Safety Information

MSDS https://www.manchesterorganics.com/E16654
Availability details for
Quantity
  • Availability
  • Add to Cart