MAN182108142 Manchester Organics

2-(3,4-Difluorophenyl)-1,3-dioxolane

  • MDL number MFCD06209062
  • Molecular Weight : 186.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L16685

Safety Information

MSDS https://www.manchesterorganics.com/L16685
Availability details for
Quantity
  • Availability
  • Add to Cart