MAN182108160 Manchester Organics

Quinoline-5-carboxylic acid

  • MDL number MFCD00047608
  • Molecular Weight : 173.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z17146

Safety Information

MSDS https://www.manchesterorganics.com/Z17146
Availability details for
Quantity
  • Availability
  • Add to Cart