MAN182108176 Manchester Organics

o-Toluic hydrazine

  • MDL number MFCD00014756
  • Cas Number : 7658-80-2
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-methylbenzohydrazide
Inchi Code 1S/C8H10N2O/c1-6-4-2-3-5-7(6)8(11)10-9/h2-5H,9H2,1H3,(H,10,11)
Inchi Key KFXLXEQCRFGDRU-UHFFFAOYSA-N

Description

Purity 98%
Country of Origin GB
Product Link https://www.manchesterorganics.com/F16680
Availability details for
Quantity
  • Availability
  • Add to Cart