MAN182108182 Manchester Organics

Ethyl 5-nitroindole-2-carboxylate

  • MDL number MFCD00216477
  • Molecular Weight : 234.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/U16976

Safety Information

MSDS https://www.manchesterorganics.com/U16976
Availability details for
Quantity
  • Availability
  • Add to Cart