MAN182108198 Manchester Organics

4-Bromo-5-phenyl-3-(trifluoromethyl)pyrazole

  • MDL number MFCD00153731
  • Molecular Weight : 291.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B16747

Safety Information

MSDS https://www.manchesterorganics.com/B16747
Availability details for
Quantity
  • Availability
  • Add to Cart