MAN182108216 Manchester Organics

3-Amino-4-bromopyridine

  • MDL number MFCD04112575
  • Molecular Weight : 173.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J18197

Safety Information

MSDS https://www.manchesterorganics.com/J18197
Availability details for
Quantity
  • Availability
  • Add to Cart