MAN182108224 Manchester Organics

3-Methyl-5-phenyl-1H-pyrazole

  • MDL number MFCD00022385
  • Molecular Weight : 158.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A10851
Availability details for
Quantity
  • Availability
  • Add to Cart