MAN182108265 Manchester Organics

4-Fluoro-3-(trifluoromethyl)benzenesulfonamide

  • MDL number MFCD10687234
  • Molecular Weight : 243.17871

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K10885

Safety Information

MSDS https://www.manchesterorganics.com/K10885
Availability details for
Quantity
  • Availability
  • Add to Cart