MAN182108267 Manchester Organics

4'-Fluoro-2'-methylacetophenone

  • MDL number MFCD00028041
  • Cas Number : 446-29-7
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/C18143
Availability details for
Quantity
  • Availability
  • Add to Cart