MAN182108274 Manchester Organics

4-Benzyloxy-3-methylbenzeneboronic acid

  • MDL number MFCD06409944
  • Cas Number : 338454-30-1
  • Molecular Weight : 242.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y16838
Availability details for
Quantity
  • Availability
  • Add to Cart