MAN182108282 Manchester Organics

6-Bromoisoquinoline

  • MDL number MFCD04973299
  • Molecular Weight : 208.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T17092
Availability details for
Quantity
  • Availability
  • Add to Cart