MAN182108323 Manchester Organics

5-Fluoro-2-nitrobenzeneboronic acid

  • MDL number MFCD09037802
  • Cas Number : 1329171-65-4
  • Molecular Weight : 184.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/F16916
Availability details for
Quantity
  • Availability
  • Add to Cart