MAN182108333 Manchester Organics

4-Isopropylbenzeneboronic acid

  • MDL number MFCD01074614
  • Molecular Weight : 164.00

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/Z17453

Safety Information

MSDS https://www.manchesterorganics.com/Z17453
Availability details for
Quantity
  • Availability
  • Add to Cart