MAN182108335 Manchester Organics

Isoquinoline-4-boronic acid

  • MDL number MFCD03412093
  • Molecular Weight : 172.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C16984

Safety Information

MSDS https://www.manchesterorganics.com/C16984
Availability details for
Quantity
  • Availability
  • Add to Cart