MAN182108358 Manchester Organics

2,3,4-Trifluorobenzeneboronic acid

  • MDL number MFCD01863168
  • Molecular Weight : 175.90

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N17136

Safety Information

MSDS https://www.manchesterorganics.com/N17136
Availability details for
Quantity
  • Availability
  • Add to Cart