MAN182108380 Manchester Organics

7-Cyanoindole

  • MDL number MFCD00800653
  • Cas Number : 96631-87-7
  • Molecular Weight : 142.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-indole-7-carbonitrile
Inchi Code 1S/C9H6N2/c10-6-8-3-1-2-7-4-5-11-9(7)8/h1-5,11H
Inchi Key NTUHBYLZRBVHRS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin GB
Product Link https://www.manchesterorganics.com/C17032
Availability details for
Quantity
  • Availability
  • Add to Cart