MAN182108382 Manchester Organics

5-Propyl-1H-pyrazol-3-ylamine

  • MDL number MFCD08061095
  • Molecular Weight : 125.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/V10894
Availability details for
Quantity
  • Availability
  • Add to Cart