MAN182108442 Manchester Organics

4-Benzyloxypyridine

  • MDL number MFCD08235112
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A18189

Safety Information

MSDS https://www.manchesterorganics.com/A18189
Availability details for
Quantity
  • Availability
  • Add to Cart