MAN182108460 Manchester Organics

3,4,5-Trifluorobenzenesulfonamide

  • MDL number MFCD09800668
  • Molecular Weight : 211.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W12102

Safety Information

MSDS https://www.manchesterorganics.com/W12102
Availability details for
Quantity
  • Availability
  • Add to Cart