MAN182108467 Manchester Organics

4-Isopropoxybenzaldehyde

  • MDL number MFCD00052357
  • Cas Number : 18962-05-5
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/M16544
Availability details for
Quantity
  • Availability
  • Add to Cart