MAN182108475 Manchester Organics

3-Methylfuran

  • MDL number MFCD00060134
  • Molecular Weight : 82.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K17536
Availability details for
Quantity
  • Availability
  • Add to Cart