MAN182108478 Manchester Organics

2-Methylfuran-3-carbonyl chloride

  • MDL number MFCD03421418
  • Molecular Weight : 144.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L17631

Safety Information

MSDS https://www.manchesterorganics.com/L17631
Availability details for
Quantity
  • Availability
  • Add to Cart