MAN182108489 Manchester Organics

2-(Chloromethyl)pyrimidine hydrochloride

  • MDL number MFCD09834372
  • Cas Number : 936643-80-0
  • Molecular Weight : 165.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/L17111
Availability details for
Quantity
  • Availability
  • Add to Cart