MAN182108508 Manchester Organics

2,5-Difluoro-4-methoxybenzaldehyde

  • MDL number MFCD06246882
  • Cas Number : 879093-08-0
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/C18191
Availability details for
Quantity
  • Availability
  • Add to Cart