MAN182108511 Manchester Organics

3,4-Difluoro-2-methylbenzoic acid

  • MDL number MFCD07777153
  • Cas Number : 157652-31-8
  • Molecular Weight : 172.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin GB
Product Link https://www.manchesterorganics.com/F18194
Availability details for
Quantity
  • Availability
  • Add to Cart