MAN182108547 Manchester Organics

Cyclopropanemethylamine

  • MDL number MFCD00037147
  • Cas Number : 2516-47-4
  • Molecular Weight : 71.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/G18218
Availability details for
Quantity
  • Availability
  • Add to Cart