MAN182108552 Manchester Organics

5-Fluoro-2-(trifluoromethyl)pyridine

  • MDL number MFCD09864698
  • Cas Number : 936841-73-5
  • Molecular Weight : 165.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/L18222
Availability details for
Quantity
  • Availability
  • Add to Cart