MAN182108564 Manchester Organics

3-Bromo-2-fluoropyridine

  • MDL number MFCD04112496
  • Molecular Weight : 175.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B18261

Safety Information

MSDS https://www.manchesterorganics.com/B18261
Availability details for
Quantity
  • Availability
  • Add to Cart