MAN182108565 Manchester Organics

2,4,5,6-Tetrafluoropyrimidine

  • MDL number MFCD10687402
  • Molecular Weight : 152.04981

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P10889

Safety Information

MSDS https://www.manchesterorganics.com/P10889
Availability details for
Quantity
  • Availability
  • Add to Cart