MAN182108568 Manchester Organics

4-Fluoro-2-(trifluoromethyl)benzenesulfonamide

  • MDL number MFCD10687237
  • Cas Number : 1040687-55-5
  • Molecular Weight : 243.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/T10892
Availability details for
Quantity
  • Availability
  • Add to Cart