MAN182108624 Manchester Organics

2-Amino-3-bromo-5-phenylpyridine

  • MDL number MFCD02183569
  • Cas Number : 107351-80-4
  • Molecular Weight : 249.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/R10157
Availability details for
Quantity
  • Availability
  • Add to Cart